Dimethyl cyclopentylmalonate ≥97.0%

Leverantör: TCI

Synonyms: Cyclopentylmalonic Acid Dimethyl Ester, Dimethyl 2-cyclopentylmalonate

C1621-25G C1621-5G
TCIAC1621-25GEA 1500 SEK
TCIAC1621-25G TCIAC1621-5G
Dimethyl cyclopentylmalonate ≥97.0%
Dimethyl cyclopentylmalonate
Formel: C₁₀H₁₆O₄
MDL Number: MFCD03844781
CAS nummer: 82491-60-9

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR