1-(2-Chloroethyl)naphthalene ≥95.0%
Leverantör: TCI EUROPE
C1490-25G
C1490-5G
TCIAC1490-25GEA
5800
SEK
TCIAC1490-25G
TCIAC1490-5G
1-(2-Chloroethyl)naphthalene ≥95.0%
1-(2-Chloroethyl)naphthalene
Formel:
C₁₂H₁₁Cl MW: 190,67 g/mol Kokpunkt: 168°C |
MDL Number:
MFCD01321149 CAS nummer: 41332-02-9 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...