Chlorodimethyl(3-phenylpropyl)silane ≥97.0%

Leverantör: TCI EUROPE
Fara

C1488-25ML C1488-5ML
TCIAC1488-25MLEA 3325 SEK
TCIAC1488-25ML TCIAC1488-5ML
Chlorodimethyl(3-phenylpropyl)silane ≥97.0%
Chlorodimethyl(3-phenylpropyl)silane
Formel: C₁₁H₁₇ClSi
MW: 212,79 g/mol
Kokpunkt: 121°C
Flampunkt: 103°C
MDL Number: MFCD00049281
CAS nummer: 17146-09-7
EINECS: 241-208-3
UN: 2987
ADR: 8,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR