2-Chloroethyl chloromethyl ether ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2-(Chloromethoxy)ethyl Chloride

C1405-25G C1405-5G
TCIAC1405-25GEA 1725 SEK
TCIAC1405-25G TCIAC1405-5G
2-Chloroethyl chloromethyl ether ≥98.0%
2-Chloroethyl chloromethyl ether
Formel: C₃H₆Cl₂O
MW: 128.98 g/mol
Kokpunkt: 147°C
Storage Temperature: Kylskåp
MDL Number: MFCD00191406
CAS nummer: 1462-33-5
UN: 2929
ADR: 6.1 II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR