(Chloromethyl)isopropoxydimethylsilane ≥96.0%

Leverantör: TCI
Varning

C1370-25ML C1370-5ML
TCIAC1370-25MLEA 2700 SEK
TCIAC1370-25ML TCIAC1370-5ML
(Chloromethyl)isopropoxydimethylsilane ≥96.0%
(Chloromethyl)isopropoxydimethylsilane
Formel: C₆H₁₅ClOSi
Kokpunkt: 145°C
Flampunkt: 28°C
MDL Number: MFCD00010605
CAS nummer: 18171-11-4
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR