Methyl (E)-4-chloro-3-methoxy-2-butenoate ≥95.0%

Leverantör: TCI
Fara

Synonyms: (E)-4-Chloro-3-methoxy-2-butenoic Acid Methyl Ester, (E)-Methyl 4-chloro-3-methoxybut-2-enoate

C1340-25G C1340-5G
TCIAC1340-25GEA 1900 SEK
TCIAC1340-25G TCIAC1340-5G
Methyl (E)-4-chloro-3-methoxy-2-butenoate ≥95.0%
Methyl (E)-4-chloro-3-methoxy-2-butenoate
Formel: C₆H₉ClO₃
MW: 164.59 g/mol
MDL Number: MFCD00071562
CAS nummer: 110104-60-4
UN: 1759
ADR: 8 III

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR