4-Chloro-2,1,3-benzoxadiazole ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
4-Chlorobenzofurazan
C1337-25G
C1337-5G
TCIAC1337-25GEA
4725
SEK
TCIAC1337-25G
TCIAC1337-5G
4-Chloro-2,1,3-benzoxadiazole ≥98.0%
4-Chloro-2,1,3-benzoxadiazole
Formel:
C₆H₃ClN₂O MW: 154,55 g/mol |
MDL Number:
MFCD00191329 CAS nummer: 7116-16-7 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 82.0 to 85.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...