(2S,8aR)-(-)-(Camphorylsulphonyl)oxaziridine ≥95.0% asymmetric oxidizing reagent

Leverantör: TCI EUROPE

Synonyms: (1R,2S)-(-)-2,N-Epoxy-10,2-camphorsultam

C1327-1G C1327-5G
TCIAC1327-1GEA 655 SEK
TCIAC1327-1G TCIAC1327-5G
(2S,8aR)-(-)-(Camphorylsulphonyl)oxaziridine ≥95.0% asymmetric oxidizing reagent
(2S,8aR)-(-)-(Camphorylsulphonyl)oxaziridine
Formel: C₁₀H₁₅NO₃S
MW: 229,29 g/mol
MDL Number: MFCD00075428
CAS nummer: 104372-31-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Specific rotation [a]20/D -47.0 to -52.0 deg (C=1, Acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR