(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide ≥95.0% asymmetric oxidizing reagent
Leverantör: TCI EUROPE
Synonyms:
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide, (+)-(2R,8aS)-(Camphorylsulphonyl)oxaziridine, (1S)-(+)-(10-Camphorsulphonyl)oxaziridine
C1326-1G
C1326-5G
TCIAC1326-1GEA
855
SEK
TCIAC1326-1G
TCIAC1326-5G
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide ≥95.0% asymmetric oxidizing reagent
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide
Formel:
C₁₀H₁₅NO₃S MW: 229,29 g/mol |
MDL Number:
MFCD00075319 CAS nummer: 104322-63-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Specific rotation [a]20/D | +47.0 to +52.0 deg (C=1, Acetone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...