Methyl-4-chloro-2-methylbutyrate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Chloro-2-methylbutyric Acid Methyl Ester
C1319-25G
C1319-5G
TCIAC1319-25GEA
1950
SEK
TCIAC1319-25G
TCIAC1319-5G
Methyl-4-chloro-2-methylbutyrate ≥98.0%
Methyl-4-chloro-2-methylbutyrate
Formel:
C₆H₁₁ClO₂ MW: 150,60 g/mol Kokpunkt: 81°C |
MDL Number:
MFCD00198001 CAS nummer: 13888-03-4 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...