3-(Chloromethyl)-3-methyloxetane ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-(Chloromethyl)-2-methyl-1,3-epoxypropane
C1229-25G C1229-5G
TCIAC1229-25GEA 1900 SEK
TCIAC1229-25G TCIAC1229-5G
3-(Chloromethyl)-3-methyloxetane ≥98.0%
3-(Chloromethyl)-3-methyloxetane
Formel: C₅H₉ClO
MW: 120,58 g/mol
Kokpunkt: 88°C
MDL Number: MFCD01709552
CAS nummer: 822-48-0
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %