(S)-(+)-1-Chloro-2-methylbutane ≥98.0%

Leverantör: TCI EUROPE
Fara

C1188-10ML C1188-25ML
TCIAC1188-10MLEA 805 SEK
TCIAC1188-10ML TCIAC1188-25ML
(S)-(+)-1-Chloro-2-methylbutane ≥98.0%
(S)-(+)-1-Chloro-2-methylbutane
Formel: C₅H₁₁Cl
MW: 106,59 g/mol
Kokpunkt: 100°C
Storage Temperature: Kylskåp
MDL Number: MFCD00221494
CAS nummer: 40560-29-0
UN: 1107
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +1.3 to +2.3 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR