Butyl-2-chloroethyl ether ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(2-Chloroethoxy)butane, 2-Chloroethyl Butyl Ether
C1171-25ML C1171-5ML
TCIAC1171-25MLEA 2075 SEK
TCIAC1171-25ML TCIAC1171-5ML
Butyl-2-chloroethyl ether ≥98.0%
Butyl-2-chloroethyl ether
Formel: C₆H₁₃ClO
MW: 136,62 g/mol
Kokpunkt: 50°C
Flampunkt: 46°C
MDL Number: MFCD00059051
CAS nummer: 10503-96-5
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afcd07966828ae1b9ab9e5e0807a8e74a4f72c79&vid=afcd07966828ae1b9ab9e5e0807a8e74a4f72c79&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %