4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-(4-(2-Phenylpropan-2-yl)phenoxy)phthalonitrile, 2-[4-(3,4-Dicyanophenoxy)phenyl]-2-phenylpropane

C1163-1G C1163-5G
TCIAC1163-1GEA 1375 SEK
TCIAC1163-1G TCIAC1163-5G
4-(4-ɑ-Cumylphenoxy)phthalonitrile ≥98.0% (by HPLC, total nitrogen)
4-(4-alpha-Cumylphenoxy)phthalonitrile
Formel: C₂₃H₁₈N₂O
MW: 338,41 g/mol
MDL Number: MFCD00059048
CAS nummer: 83482-57-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 90.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR