Naftifine hydrochloride ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Naftifine HCl, (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine hydrochloride, AW 105843, SN 105843, N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine hydrochloride

C1162-1G C1162-5G
TCIAC1162-1GEA 1275 SEK
TCIAC1162-1G TCIAC1162-5G
Naftifine hydrochloride ≥98.0%
Naftifine hydrochloride
Formel: C₂₁H₂₁N
MW: 323,86 g/mol
MDL Number: MFCD00059047
CAS nummer: 65473-14-5
Merck Index: 14,06355

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 174.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR