2-(1-Cyclohexenyl)cyclohexanone ≥85.0%
Leverantör: TCI EUROPE
Synonyms: [1,1'-Bi(cyclohexan)]-1'-en-2-one
C1160-25ML C1160-500ML
TCIAC1160-25MLEA 215 SEK
TCIAC1160-25ML TCIAC1160-500ML
2-(1-Cyclohexenyl)cyclohexanone ≥85.0%
2-(1-Cyclohexenyl)cyclohexanone
Formel: C₁₂H₁₈O
MW: 178,28 g/mol
Kokpunkt: 275°C
MDL Number: MFCD00013796
CAS nummer: 1502-22-3
EINECS: 216-120-3
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 85.0 %