Sodium-3-chloro-2-hydroxypropanesulphonate ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
3-Chloro-2-hydroxypropanesulphonic acid Sodium Salt, Sodium 3-chloro-2-hydroxypropane-1-sulphonate
C1144-25G
C1144-500G
TCIAC1144-25GEA
215
SEK
TCIAC1144-25G
TCIAC1144-500G
Sodium-3-chloro-2-hydroxypropanesulphonate ≥95.0%
Sodium-3-chloro-2-hydroxypropanesulphonate
Formel:
C₃H₇ClO₄S MW: 196,58 g/mol |
MDL Number:
MFCD00013378 CAS nummer: 126-83-0 EINECS: 204-807-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...