3-Chloropropylamine hydrochloride ≥97.0%

Leverantör: TCI
Varning

Synonyms: 3-Chloropropan-1-amine hydrochloride

C1062-25G C1062-100G
TCIAC1062-25GEA 215 SEK
TCIAC1062-25G TCIAC1062-100G
3-Chloropropylamine hydrochloride ≥97.0%
3-Chloropropylamine hydrochloride
Formel: C₃H₈ClN
MW: 130.01 g/mol
MDL Number: MFCD00012913
CAS nummer: 6276-54-6
EINECS: 228-474-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=009cf55dc674e088072d45bc3015b6e2cabbd23e&vid=009cf55dc674e088072d45bc3015b6e2cabbd23e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 147.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR