N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: N-(2,3-Xylyl)-N-cinnamoylhydroxylamine

C1058-1G C1058-5G
TCIAC1058-1GEA 685 SEK
TCIAC1058-1G TCIAC1058-5G
N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine ≥98.0% (by HPLC, total nitrogen)
N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine
Formel: C₁₇H₁₇NO₂
MW: 267,33 g/mol
MDL Number: MFCD00059308
CAS nummer: 69891-38-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Suitability for vanadium-analysis to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR