Vinyl-2-chlorobenzoate ≥98.0% stabilised

Leverantör: TCI
Varning


Stabilization: med hydrokinon

C1048-25G C1048-5G
TCIAC1048-25GEA 5475 SEK
TCIAC1048-25G TCIAC1048-5G
Vinyl-2-chlorobenzoate ≥98.0% stabilised
Vinyl-2-chlorobenzoate
Formel: C₉H₇ClO₂
Kokpunkt: 130°C
MDL Number: MFCD00059479
CAS nummer: 15721-27-4

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR