Propyl cyanoacetate ≥99.0%

Leverantör: TCI
Varning

Synonyms: Propyl 2-cyanoacetate, Cyanoacetic Acid Propyl Ester

C1016-25ML C1016-500ML
TCIAC1016-25MLEA 295 SEK
TCIAC1016-25ML TCIAC1016-500ML
Propyl cyanoacetate ≥99.0%
Propyl cyanoacetate
Formel: C₆H₉NO₂
Kokpunkt: 216°C
Flampunkt: 108°C
MDL Number: MFCD00015784
CAS nummer: 14447-15-5
EINECS: 238-424-5

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR