Ethyl-3-cyclohexene-1-carboxylate ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 3-Cyclohexene-1-carboxylic Acid Ethyl Ester
C0966-25ML C0966-5ML
TCIAC0966-25MLEA 2250 SEK
TCIAC0966-25ML TCIAC0966-5ML
Ethyl-3-cyclohexene-1-carboxylate ≥95.0%
Ethyl-3-cyclohexene-1-carboxylate
Formel: C₉H₁₄O₂
MW: 154,21 g/mol
Kokpunkt: 195°C
MDL Number: MFCD00037158
CAS nummer: 15111-56-5
EINECS: 239-161-9
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %