Vinyl cinnamate ≥99.0% stabilised

Leverantör: TCI EUROPE

Synonyms: Cinnamic Acid Vinyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

C0899-25ML C0899-500ML
TCIAC0899-25MLEA 535 SEK
TCIAC0899-25ML TCIAC0899-500ML
Vinyl cinnamate ≥99.0% stabilised
Vinyl cinnamate
Formel: C₁₁H₁₀O₂
MW: 174,20 g/mol
Kokpunkt: 125°C
Flampunkt: 132°C
MDL Number: MFCD00059324
CAS nummer: 3098-92-8

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR