Vinyl hexanoate ≥99.0% stabilised

Leverantör: TCI
Varning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

C0898-25ML C0898-500ML
TCIAC0898-25MLEA 285 SEK
TCIAC0898-25ML TCIAC0898-500ML
Vinyl hexanoate ≥99.0% stabilised
Vinyl hexanoate
Formel: C₈H₁₄O₂
Kokpunkt: 167°C
Flampunkt: 51°C
MDL Number: MFCD00048869
CAS nummer: 3050-69-9
EINECS: 221-265-0
UN: 3272
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR