1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥98.0%

Leverantör: TCI EUROPE
Varning

C0857-25G C0857-5G
TCIAC0857-25GEA 4775 SEK
TCIAC0857-25G TCIAC0857-5G
1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥98.0%
1,2,3,4-Cyclopentanetetracarboxylic dianhydride
Formel: C₉H₆O₆
MW: 210,14 g/mol
MDL Number: MFCD00046872
CAS nummer: 6053-68-5
EINECS: 227-964-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e28f5ee82d3eafb6668bae1b55716a610b7f54da&vid=e28f5ee82d3eafb6668bae1b55716a610b7f54da&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR