1,2,3,4-Cyclopentanetetracarboxylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

C0856-25G C0856-5G
TCIAC0856-25GEA 1525 SEK
TCIAC0856-25G TCIAC0856-5G
1,2,3,4-Cyclopentanetetracarboxylic acid ≥98.0% (by GC, titration analysis)
1,2,3,4-Cyclopentanetetracarboxylic acid
Formel: C₉H₁₀O₈
MW: 246.17 g/mol
MDL Number: MFCD00001377
CAS nummer: 3724-52-5
EINECS: 223-074-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR