4-Chlorophenoxyacetyl chloride ≥98.0%
Leverantör: TCI
|
Fara
|
C0832-25G
C0832-500G
C0832-100G
TCIAC0832-25GEA
735
SEK
TCIAC0832-25G
TCIAC0832-500G
TCIAC0832-100G
4-Chlorophenoxyacetyl chloride ≥98.0%
4-Chlorophenoxyacetyl chloride
Formel:
C₈H₆Cl₂O₂ Kokpunkt: 165°C |
MDL Number:
MFCD00000727 CAS nummer: 4122-68-3 EINECS: 223-923-2 UN: 3265 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Freezing point | 15.0 to 23.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...