N-Cyclohexyl-2-benzothiazolylsulphenamide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
C0576-25G
C0576-500G
TCIAC0576-25GEA
245
SEK
TCIAC0576-25G
TCIAC0576-500G
N-Cyclohexyl-2-benzothiazolylsulphenamide ≥98.0% (by HPLC, total nitrogen)
N-Cyclohexyl-2-benzothiazolylsulphenamide
Formel:
C₁₃H₁₆N₂S₂ |
MDL Number:
MFCD00022872 CAS nummer: 95-33-0 EINECS: 202-411-2 UN: 3077 ADR: 9,III |
|
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 98.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...