trans-Cinnamamide ≥98.0% (by GC, total nitrogen)

Leverantör: TCI
Varning

C0536-25G C0536-5G
TCIAC0536-25GEA 1325 SEK
TCIAC0536-25G TCIAC0536-5G
trans-Cinnamamide ≥98.0% (by GC, total nitrogen)
trans-Cinnamamide
Formel: C₉H₉NO
MDL Number: MFCD00008033
CAS nummer: 22031-64-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 147.0 to 151.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR