Chrysophenine

Leverantör: TCI EUROPE

Synonyms: Chrysophenine sodium salt

C0530-25G C0530-500G
TCIAC0530-25GEA 285 SEK
TCIAC0530-25G TCIAC0530-500G
Chrysophenine
Chrysophenine
Formel: C₃₀H₂₈N₄O₈S₂
MDL Number: MFCD00007488
CAS nummer: 2870-32-8
EINECS: 220-698-2

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Absorbance (E1%1cm) min. 500 (395.0 to 400.0 nm、H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR