Cytidine-5'-diphosphate trisodium salt hydrate ≥95.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: CDP trisodium salt, 5'-CDP 3Na

C0523-1G C0523-10MG C0523-100MG
TCIAC0523-1GEA 2225 SEK
TCIAC0523-1G TCIAC0523-10MG TCIAC0523-100MG
Cytidine-5'-diphosphate trisodium salt hydrate ≥95.0% (by HPLC, titration analysis)
Cytidine-5'-diphosphate trisodium salt hydrate
Formel: C₉H₁₅N₃O₁₁P₂·H₂O
MW: 469,12 g/mol (anhy)
Storage Temperature: Kylskåp
MDL Number: MFCD00069770
CAS nummer: 34393-59-4
EINECS: 251-990-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) 95.0 %以上 (無水物換算)
Water max. 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR