Cyclohexylamine hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Cyclohexylamine Hydrochloride

C0495-25G C0495-500G
TCIAC0495-25GEA 230 SEK
TCIAC0495-25G TCIAC0495-500G
Cyclohexylamine hydrochloride ≥98.0%
Cyclohexylamine hydrochloride
Formel: C₆H₁₃N
MW: 135,64 g/mol
MDL Number: MFCD00050540
CAS nummer: 4998-76-9
EINECS: 225-661-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR