Cryptocyanine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,1'-Diethyl-4,4'-carbocyanine Iodide

C0426-1G C0426-200MG
TCIAC0426-1GEA 1600 SEK
TCIAC0426-1G TCIAC0426-200MG
Cryptocyanine ≥98.0%
Cryptocyanine
Formel: C₂₅H₂₅IN₂
MW: 480,39 g/mol
MDL Number: MFCD00011970
CAS nummer: 4727-50-8
EINECS: 225-224-8
UN: 2811
ADR: 6.1,II

Order Now

Specifikation testresultat

Appearance Yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR