Butyl crotonate ≥97.0%

Leverantör: TCI EUROPE

Synonyms: Crotonic Acid Butyl Ester

C0417-25ML C0417-500ML
TCIAC0417-25MLEA 375 SEK
TCIAC0417-25ML TCIAC0417-500ML
Butyl crotonate ≥97.0%
Butyl crotonate
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Kokpunkt: 180°C
Flampunkt: 64°C
MDL Number: MFCD00053794
CAS nummer: 7299-91-4
EINECS: 230-742-2

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR