Sodium-4-chlorobenzenesulphonate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Chlorobenzenesulphonic acid Sodium Salt

C0127-25G C0127-500G
TCIAC0127-25GEA 775 SEK
TCIAC0127-25G TCIAC0127-500G
Sodium-4-chlorobenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-chlorobenzenesulphonate
Formel: C₆H₅ClO₃S
MW: 214,59 g/mol
MDL Number: MFCD00014731
CAS nummer: 5138-90-9
EINECS: 225-900-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR