1-Bromo-2-methoxynaphthalene ≥98.0%

Leverantör: TCI EUROPE
Varning

B6103-25G B6103-5G
TCIAB6103-25GEA 1975 SEK
TCIAB6103-25G TCIAB6103-5G
1-Bromo-2-methoxynaphthalene ≥98.0%
1-Bromo-2-methoxynaphthalene
Formel: C₁₁H₉BrO
MW: 237,1 g/mol
Kokpunkt: 192°C
MDL Number: MFCD00055374
CAS nummer: 3401-47-6
EINECS: 222-272-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR