1,4-Bis(dimethylvinylsilyl)benzene ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,4-Bis(dimethyl(vinyl)silyl)benzene, 1,4-Phenylenebis[dimethyl(vinyl)silane]

B6054-25G B6054-5G
TCIAB6054-25GEA 3375 SEK
TCIAB6054-25G TCIAB6054-5G
1,4-Bis(dimethylvinylsilyl)benzene ≥95.0%
1,4-Bis(dimethylvinylsilyl)benzene
Formel: C₁₄H₂₂Si₂
MW: 246,50 g/mol
Kokpunkt: 95°C
MDL Number: MFCD07778038
CAS nummer: 4519-17-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR