1-Butyl-1-methylpyrrolidiniumhexafluorophosphate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-Butyl-N-methylpyrrolidinium hexafluorophosphate

B6039-5G B6039-25G
TCIAB6039-5GEA 565 SEK
TCIAB6039-5G TCIAB6039-25G
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate ≥98.0%
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate
Formel: C₉H₂₀F₆NP
MW: 287,23 g/mol
MDL Number: MFCD08458501
CAS nummer: 330671-29-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR