3-Bromophenanthrene-9,10-dione ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Bromo-9,10-phenanthrenedione

B6037-1G B6037-5G
TCIAB6037-1GEA 590 SEK
TCIAB6037-1G TCIAB6037-5G
3-Bromophenanthrene-9,10-dione ≥95.0%
3-Bromophenanthrene-9,10-dione
Formel: C₁₄H₇BrO₂
MW: 287,11 g/mol
MDL Number: MFCD18072876
CAS nummer: 13292-05-2

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR