N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
(S)-TERT-BUTYL 1-(METHOXY(METHYL)AMINO)-1-OXOPROPAN-2-YLCARBAMATE, BOC-L-ALANINE N-METHOXY-N-METHYL AMIDE, (2S)-2-[(tert-butoxy)carbonylamino]-N-methoxy-N-methylpropanamide, Boc-Ala-NMe(OMe)
B6019-1G
B6019-5G
TCIAB6019-1GEA
415
SEK
TCIAB6019-1G
TCIAB6019-5G
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide ≥98.0%
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide
Formel:
C₁₀H₂₀N₂O₄ MW: 232,28 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00151859 CAS nummer: 87694-49-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Specific rotation [a]20/D | -27.0 to -31.0 deg (C=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...