2,4-Bis[(dodecylthio)methyl]-6-methylphenol ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 2,4-Bis(dodecylsulphanylmethyl)-6-methylphenol
B6016-25G B6016-500G
TCIAB6016-25GEA 325 SEK
TCIAB6016-25G TCIAB6016-500G
2,4-Bis[(dodecylthio)methyl]-6-methylphenol ≥95.0%
2,4-Bis[(dodecylthio)methyl]-6-methylphenol
Formel: C₃₃H₆₀OS₂
MW: 536,96 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11973660
CAS nummer: 110675-26-8
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %