Buformin hydrochloride ≥95.0%

Leverantör: TCI
Varning

B6007-50MG B6007-250MG
TCIAB6007-50MGEA 525 SEK
TCIAB6007-50MG TCIAB6007-250MG
Buformin hydrochloride ≥95.0%
Buformin hydrochloride
Formel: C₆H₁₅N₅·HCl
Storage Temperature: Frys
MDL Number: MFCD00045591
CAS nummer: 1190-53-0
EINECS: 214-723-6
Merck Index: 14,01476

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 174.0 to 178.0 deg-C
Elemental analysis (Nitrogen) 34.30 to 38.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR