1,3,5-Triformylbenzene ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1,3,5-benzenetricarbaldehyde, Trimesic formaldehyde, Benzene-1,3,5-tricarbaldehyde
B6003-200MG B6003-1G
TCIAB6003-200MGEA 495 SEK
TCIAB6003-200MG TCIAB6003-1G
1,3,5-Triformylbenzene ≥98.0%
1,3,5-Triformylbenzene
Formel: C₉H₆O₃
MW: 162,14 g/mol
MDL Number: MFCD00182470
CAS nummer: 3163-76-6
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C