1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate) ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
DCyPB-FB
B5991-1G
B5991-5G
TCIAB5991-1GEA
965
SEK
TCIAB5991-1G
TCIAB5991-5G
1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate) ≥95.0%
1,4-Bis(dicyclohexylphosphonium)butane bis(tetrafluoroborate)
Formel:
C₂₈H₅₄B₂F₈P₂ |
MDL Number:
MFCD22666031 CAS nummer: 1389309-54-9 UN: 1759 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Melting point | 247.0 to 251.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...