2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone ≥98.0%
Leverantör: TCI
B5987-1G
B5987-5G
TCIAB5987-1GEA
910
SEK
TCIAB5987-1G
TCIAB5987-5G
2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone ≥98.0%
2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone
Formel:
C₂₉H₁₈Br₂O MW: 542.27 g/mol |
CAS nummer:
54523-24-9 |
Specifikation testresultat
Appearance | Red to Dark blue to Black powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 247.0 to 251.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...