Bis[4-(tert-butyl)phenyl]iodonium tetra(nonafluoro-tert-butoxy)aluminate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Bis[4-(tert-butyl)phenyl]iodonium tetra(perfluoro-tert-butoxy)aluminate, Bis[4-(tert-butyl)phenyl]iodonium tetrakis[[1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-yl]oxy]aluminate

B5956-100MG B5956-1G
TCIAB5956-100MGEA 1400 SEK
TCIAB5956-100MG TCIAB5956-1G
Bis[4-(tert-butyl)phenyl]iodonium tetra(nonafluoro-tert-butoxy)aluminate ≥98.0% (by HPLC, titration analysis)
Bis[4-(tert-butyl)phenyl]iodonium tetra(nonafluoro-tert-butoxy)aluminate
Formel: C₃₆H₂₆AlF₃₆IO₄
MW: 1360,43 g/mol
CAS nummer: 2350272-68-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR