1-Bromo-3,3-dimethylbutane ≥98.0%
Leverantör: TCI EUROPE
Varning
B5948-25G B5948-5G
TCIAB5948-25GEA 2500 SEK
TCIAB5948-25G TCIAB5948-5G
1-Bromo-3,3-dimethylbutane ≥98.0%
1-Bromo-3,3-dimethylbutane
Formel: C₆H₁₃Br
MW: 165,07 g/mol
Kokpunkt: 138°C
MDL Number: MFCD00060844
CAS nummer: 1647-23-0
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %