2-Bromo-5-tert-butyl-1,3-dimethylbenzene ≥98.0%

Leverantör: TCI

B5924-1G B5924-5G
TCIAB5924-1GEA 975 SEK
TCIAB5924-1G TCIAB5924-5G
2-Bromo-5-tert-butyl-1,3-dimethylbenzene ≥98.0%
2-Bromo-5-tert-butyl-1,3-dimethylbenzene
Formel: C₁₂H₁₇Br
MDL Number: MFCD12973868
CAS nummer: 5345-05-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR