4-Bromooxindole ≥98.0%
Leverantör: TCI
Synonyms:
4-Bromooxindole, 4-Bromo-2,3-dihydro-1H-indol-2-one, 4-Bromoindolin-2-one
B5903-1G
B5903-200MG
TCIAB5903-1GEA
1725
SEK
TCIAB5903-1G
TCIAB5903-200MG
4-Bromooxindole ≥98.0%
4-Bromooxindole
Formel:
C₈H₆BrNO MW: 212.05 g/mol |
MDL Number:
MFCD06659911 CAS nummer: 99365-48-7 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...