tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid tert-Butyl Ester, NH2-PEG4-(CH2)2COOtBu, Amino-PEG4-Boc

B5900-250MG B5900-1G
TCIAB5900-250MGEA 765 SEK
TCIAB5900-250MG TCIAB5900-1G
tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate ≥95.0%
tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate
Formel: C₁₅H₃₁NO₆
MW: 321,41 g/mol
Storage Temperature: Frys
MDL Number: MFCD11041116
CAS nummer: 581065-95-4

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR